Assay Details
| Gene Symbol: Stradb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172656.5 | NP_766244.4 | 10 - 11 | 1448 | 80 | |
| GenBank mRNA | AB057666.1 | - | 10 - 11 | 1215 | 80 | |
| AB057667.1 | - | 9 - 10 | 1043 | 80 | ||
| AK080096.1 | - | 10 - 11 | 1412 | 80 | ||
| AK140272.1 | - | 9 - 10 | 1326 | 80 | ||
| AK146079.1 | - | 10 - 11 | 1261 | 80 | ||
| BC037003.1 | - | 10 - 11 | 1355 | 80 | ||
| BC055814.1 | - | 11 - 12 | 1588 | 80 | ||
| BC060956.1 | - | 10 - 11 | 1305 | 30316511 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stradb
Genomic Map