Assay Details
| Gene Symbol: Stradb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172656.5 | NP_766244.4 | 4 - 5 | 572 | 70 | |
| GenBank mRNA | AB057666.1 | - | 4 - 5 | 339 | 70 | |
| AB057667.1 | - | 4 - 5 | 339 | 70 | ||
| AK046790.1 | - | 545 | 70 | |||
| AK080096.1 | - | 4 - 5 | 536 | 70 | ||
| AK086974.1 | - | 4 - 5 | 503 | 70 | ||
| AK140272.1 | - | 4 - 5 | 622 | 70 | ||
| AK146079.1 | - | 4 - 5 | 385 | 70 | ||
| BC037003.1 | - | 4 - 5 | 481 | 70 | ||
| BC055814.1 | - | 5 - 6 | 712 | 70 | ||
| BC060956.1 | - | 4 - 5 | 429 | 30316511 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stradb
Genomic Map