Assay Details
| Gene Symbol: Stradb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172656.5 | NP_766244.4 | 7 - 8 | 922 | 78 | |
| GenBank mRNA | AB057666.1 | - | 7 - 8 | 689 | 78 | |
| AK046790.1 | - | 895 | 78 | |||
| AK080096.1 | - | 7 - 8 | 886 | 78 | ||
| AK146079.1 | - | 7 - 8 | 735 | 78 | ||
| BC037003.1 | - | 7 - 8 | 831 | 78 | ||
| BC055814.1 | - | 8 - 9 | 1062 | 78 | ||
| BC060956.1 | - | 7 - 8 | 779 | 30316511 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stradb
Genomic Map