Assay Details
| Gene Symbol: Stradb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172656.5 | NP_766244.4 | 8 - 9 | 1093 | 84 | |
| GenBank mRNA | AB057666.1 | - | 8 - 9 | 860 | 84 | |
| AK044340.1 | - | 1 - 2 | 950 | 84 | ||
| AK046790.1 | - | 1066 | 84 | |||
| AK080096.1 | - | 8 - 9 | 1057 | 84 | ||
| AK146079.1 | - | 8 - 9 | 906 | 84 | ||
| BC055814.1 | - | 9 - 10 | 1233 | 84 | ||
| BC060956.1 | - | 8 - 9 | 950 | 30316511 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stradb
Genomic Map