Assay Details
| Gene Symbol: Stradb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172656.5 | NP_766244.4 | 9 - 10 | 1192 | 67 | |
| GenBank mRNA | AB057666.1 | - | 9 - 10 | 959 | 67 | |
| AB057667.1 | - | 8 - 9 | 787 | 67 | ||
| AK044340.1 | - | 2 - 3 | 1049 | 67 | ||
| AK046790.1 | - | 1165 | 67 | |||
| AK080096.1 | - | 9 - 10 | 1156 | 67 | ||
| AK140272.1 | - | 8 - 9 | 1070 | 67 | ||
| AK146079.1 | - | 9 - 10 | 1005 | 67 | ||
| BC037003.1 | - | 9 - 10 | 1099 | 67 | ||
| BC055814.1 | - | 10 - 11 | 1332 | 67 | ||
| BC060956.1 | - | 9 - 10 | 1049 | 30316511 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stradb
Genomic Map