Assay Details
| Gene Symbol: Casp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080126.1 | NP_001073595.1 | 7 - 8 | 1029 | 122 | |
| NM_001277926.1 | NP_001264855.1 | 8 - 9 | 1145 | 122 | ||
| NM_009812.2 | NP_033942.1 | 7 - 8 | 1202 | 122 | ||
| GenBank mRNA | AF067834.1 | - | 7 - 8 | 1082 | 122 | |
| AJ000641.1 | - | 6 - 7 | 648 | 122 | ||
| AJ007749.1 | - | 7 - 8 | 855 | 122 | ||
| AK146758.1 | - | 7 - 8 | 1202 | 122 | ||
| AK153340.1 | - | 7 - 8 | 911 | 122 | ||
| AK156135.1 | - | 7 - 8 | 918 | 122 | ||
| AK168812.1 | - | 7 - 8 | 1202 | 122 | ||
| BC006737.1 | - | 7 - 8 | 1144 | 3964119 | 122 | |
| BC049955.1 | - | 7 - 8 | 1036 | 4208824 | 122 | |
| CT010166.1 | - | 6 - 7 | 809 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp8
Genomic Map