| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400033071.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033071.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033071.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033072.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033072.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033072.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033070.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033070.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033070.mm.1 |
NM_011122.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033075.mm.1 |
TC0400004004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400033075.mm.1 |
NM_011122.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033075.mm.1 |
NM_011122.3 |
Refseq |