Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 11 - 12 | 1297 | 71 | |
| GenBank mRNA | AK137480.1 | - | 11 - 12 | 1248 | 71 | |
| BC006599.1 | - | 11 - 12 | 1222 | 3965301 | 71 | |
| BC010268.1 | - | 3 - 4 | 242 | 3589182 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Plod1
Genomic Map