Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 14 - 15 | 1679 | 89 | |
| GenBank mRNA | AF046782.1 | - | 14 - 15 | 1607 | 89 | |
| AK137480.1 | - | 14 - 15 | 1630 | 89 | ||
| AK213092.1 | - | 1 - 2 | 83 | 89 | ||
| BC006599.1 | - | 14 - 15 | 1604 | 3965301 | 89 | |
| BC010268.1 | - | 6 - 7 | 624 | 3589182 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Plod1
Genomic Map