Assay Details
| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 16 - 17 | 1855 | 78 | |
| GenBank mRNA | AF046782.1 | - | 16 - 17 | 1783 | 78 | |
| AK137480.1 | - | 16 - 17 | 1806 | 78 | ||
| BC006599.1 | - | 16 - 17 | 1780 | 3965301 | 78 | |
| BC010268.1 | - | 8 - 9 | 800 | 3589182 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod1
Genomic Map