Assay Details
| Gene Symbol: Acsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033606.2 | NP_001028778.2 | 10 - 11 | 1780 | 73 | |
| NM_001136222.1 | NP_001129694.1 | 9 - 10 | 1362 | 73 | ||
| NM_028817.3 | NP_083093.2 | 11 - 12 | 1920 | 73 | ||
| GenBank mRNA | AK012088.1 | - | 10 - 11 | 1723 | 73 | |
| AK160839.1 | - | 9 - 10 | 1306 | 73 | ||
| AK161708.1 | - | 11 - 12 | 1921 | 73 | ||
| AK166723.1 | - | 11 - 12 | 1858 | 73 | ||
| BC031529.1 | - | 9 - 10 | 1505 | 4009004 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl3
Genomic Map