Assay Details
| Gene Symbol: Acsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033606.2 | NP_001028778.2 | 12 - 13 | 2047 | 63 | |
| NM_001136222.1 | NP_001129694.1 | 11 - 12 | 1629 | 63 | ||
| NM_028817.3 | NP_083093.2 | 13 - 14 | 2187 | 63 | ||
| GenBank mRNA | AK012088.1 | - | 12 - 13 | 1990 | 63 | |
| AK160839.1 | - | 11 - 12 | 1573 | 63 | ||
| AK161708.1 | - | 13 - 14 | 2188 | 63 | ||
| AK166723.1 | - | 13 - 14 | 2125 | 63 | ||
| BC031529.1 | - | 11 - 12 | 1772 | 4009004 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl3
Genomic Map