| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100005855.mm.1 |
NM_001001981.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005855.mm.1 |
NM_001001981.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005855.mm.1 |
NM_001136226.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005855.mm.1 |
NM_001136226.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005855.mm.1 |
NM_028817.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005855.mm.1 |
NM_028817.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005855.mm.1 |
TC0100000682.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005854.mm.1 |
NM_001001981.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005854.mm.1 |
NM_001001981.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005854.mm.1 |
NM_001136226.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005854.mm.1 |
NM_001136226.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005854.mm.1 |
NM_028817.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005854.mm.1 |
NM_028817.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005854.mm.1 |
TC0100000682.mm.1 |
MTA TC |