Assay Details
| Gene Symbol: Pax3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159520.1 | NP_001152992.1 | 6 - 7 | 1345 | 66 | |
| NM_008781.4 | NP_032807.3 | 6 - 7 | 1345 | 66 | ||
| GenBank mRNA | AK012493.1 | - | 6 - 7 | 1047 | 66 | |
| AK014337.1 | - | 6 - 7 | 1047 | 66 | ||
| AK044985.1 | - | 6 - 7 | 1050 | 66 | ||
| AK045018.1 | - | 6 - 7 | 1346 | 66 | ||
| AK144973.1 | - | 6 - 7 | 1050 | 66 | ||
| AK147919.1 | - | 6 - 7 | 1050 | 66 | ||
| AK148354.1 | - | 6 - 7 | 1326 | 66 | ||
| AK157760.1 | - | 6 - 7 | 1047 | 66 | ||
| BC048699.1 | - | 6 - 7 | 1016 | 6518115 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pax3
Genomic Map