Assay Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278075.1 | NP_001265004.1 | 5 - 6 | 508 | 79 | |
| NM_011811.4 | NP_035941.2 | 5 - 6 | 508 | 79 | ||
| NR_102757.1 | 5 - 6 | 410 | 79 | |||
| NR_102758.1 | 4 - 5 | 452 | 79 | |||
| GenBank mRNA | AF123263.1 | - | 5 - 6 | 508 | 79 | |
| AK010271.1 | - | 5 - 6 | 471 | 79 | ||
| AK012154.1 | - | 5 - 6 | 471 | 79 | ||
| AK045432.1 | - | 5 - 6 | 460 | 79 | ||
| AK076607.1 | - | 5 - 6 | 470 | 79 | ||
| AK134799.1 | - | 4 - 5 | 414 | 79 | ||
| AK165645.1 | - | 5 - 6 | 375 | 79 | ||
| AK167945.1 | - | 5 - 6 | 471 | 79 | ||
| AK168923.1 | - | 5 - 6 | 452 | 79 | ||
| AK169621.1 | - | 4 - 5 | 415 | 79 | ||
| BC002313.1 | - | 5 - 6 | 367 | 3592805 | 79 | |
| BC016428.1 | - | 5 - 6 | 475 | 4239906 | 79 | |
| CN693684.1 | - | 2 - 3 | 196 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Farsb
Genomic Map