Assay Details
| Gene Symbol: Ctss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267695.2 | NP_001254624.2 | 2 - 3 | 272 | 151 | |
| NM_021281.3 | NP_067256.4 | 2 - 3 | 269 | 151 | ||
| GenBank mRNA | AF038546.1 | - | 2 - 3 | 211 | 151 | |
| AK028366.1 | - | 2 - 3 | 271 | 151 | ||
| AK150274.1 | - | 2 - 3 | 203 | 151 | ||
| AK150758.1 | - | 1 - 2 | 115 | 151 | ||
| AK150842.1 | - | 1 - 2 | 115 | 151 | ||
| AK152191.1 | - | 2 - 3 | 222 | 151 | ||
| AK152618.1 | - | 1 - 2 | 115 | 151 | ||
More Information
Set Membership: |
|

Ctss
Genomic Map