Assay Details
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037737.2 | NP_001032826.1 | 18 - 19 | 1956 | 86 | |
| NM_009709.4 | NP_033839.2 | 17 - 18 | 1911 | 86 | ||
| GenBank mRNA | AK028546.1 | - | 17 - 18 | 1898 | 86 | |
| AK040475.1 | - | 18 - 19 | 1958 | 86 | ||
| AK145415.1 | - | 17 - 18 | 1866 | 86 | ||
| AK152473.1 | - | 18 - 19 | 1938 | 86 | ||
| AK153355.1 | - | 18 - 19 | 1938 | 86 | ||
| AK220458.1 | - | 14 - 15 | 2087 | 86 | ||
| BC012870.1 | - | 18 - 19 | 1918 | 3964932 | 86 | |
| U10325.1 | - | 18 - 19 | 1820 | 86 | ||
| U14333.1 | - | 17 - 18 | 1749 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arnt
Genomic Map