Assay Details
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037737.2 | NP_001032826.1 | 7 - 8 | 851 | 86 | |
| NM_009709.4 | NP_033839.2 | 6 - 7 | 806 | 86 | ||
| GenBank mRNA | AK028546.1 | - | 6 - 7 | 793 | 86 | |
| AK040475.1 | - | 7 - 8 | 853 | 86 | ||
| AK141362.1 | - | 7 - 8 | 722 | 86 | ||
| AK145415.1 | - | 6 - 7 | 761 | 86 | ||
| AK152473.1 | - | 7 - 8 | 833 | 86 | ||
| AK153355.1 | - | 7 - 8 | 833 | 86 | ||
| AK220458.1 | - | 3 - 4 | 982 | 86 | ||
| BC012870.1 | - | 7 - 8 | 813 | 3964932 | 86 | |
| U10325.1 | - | 7 - 8 | 715 | 86 | ||
| U14333.1 | - | 6 - 7 | 644 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arnt
Genomic Map