Assay Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033302.2 | NP_001028474.1 | 1 - 2 | 79 | 83 | |
| GenBank mRNA | AK164001.1 | - | 1 - 2 | 79 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164001.1, NM_001033302.2 transcripts.
Set Membership: |
|

Ciart
Genomic Map