Assay Details
| Gene Symbol: Prpf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027541.4 | NP_081817.2 | 11 - 12 | 2012 | 82 | |
| GenBank mRNA | AK014398.1 | - | 11 - 12 | 1620 | 82 | |
| AK088194.1 | - | 11 - 12 | 2013 | 82 | ||
| AK139852.1 | - | 11 - 12 | 1645 | 82 | ||
| AK167435.1 | - | 11 - 12 | 1649 | 82 | ||
| BC006782.1 | - | 11 - 12 | 1597 | 3589226 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prpf3
Genomic Map