Assay Details
| Gene Symbol: Prpf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK139852.1 | - | 1 - 2 | 67 | 68 | |
| AK167435.1 | - | 1 - 2 | 71 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139852.1,AK167435.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prpf3
Genomic Map