Assay Details
| Gene Symbol: Me2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145494.2 | NP_663469.1 | 5 - 6 | 569 | 65 | |
| GenBank mRNA | AK042289.1 | - | 5 - 6 | 616 | 65 | |
| AK043103.1 | - | 5 - 6 | 606 | 65 | ||
| AK050933.1 | - | 5 - 6 | 616 | 65 | ||
| AK050980.1 | - | 5 - 6 | 616 | 65 | ||
| AK077746.1 | - | 5 - 6 | 616 | 65 | ||
| AK156403.1 | - | 5 - 6 | 616 | 65 | ||
| AK162126.1 | - | 5 - 6 | 628 | 65 | ||
| AK171157.1 | - | 5 - 6 | 616 | 65 | ||
| BC004709.1 | - | 5 - 6 | 569 | 3499365 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Me2
Genomic Map