Assay Details
| Gene Symbol: Pdzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033222.3 | NP_001028394.2 | 1 - 2 | 1062 | 62 | |
| GenBank mRNA | AK041847.1 | - | 1 - 2 | 516 | 62 | |
| AK145020.1 | - | 1 - 2 | 1062 | 62 | ||
| BC141370.1 | - | 1 - 2 | 967 | 9055983 | 62 | |
| BC141371.1 | - | 1 - 2 | 967 | 9055984 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdzd8
Genomic Map