Assay Details
| Gene Symbol: Pdzd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033222.3 | NP_001028394.2 | 2 - 3 | 1189 | 93 | |
| GenBank mRNA | AK041847.1 | - | 2 - 3 | 643 | 93 | |
| AK145020.1 | - | 2 - 3 | 1189 | 93 | ||
| BC141370.1 | - | 2 - 3 | 1094 | 9055983 | 93 | |
| BC141371.1 | - | 2 - 3 | 1094 | 9055984 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdzd8
Genomic Map