Assay Details
| Gene Symbol: Ift74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290568.1 | NP_001277497.1 | 12 - 13 | 1144 | 137 | |
| NM_026319.3 | NP_080595.2 | 12 - 13 | 1144 | 137 | ||
| GenBank mRNA | AK006492.1 | - | 3 - 4 | 251 | 137 | |
| AK053369.1 | - | 12 - 13 | 1092 | 137 | ||
| BY706530.1 | - | 3 - 4 | 249 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ift74
Genomic Map