Assay Details
| Gene Symbol: Ift74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290568.1 | NP_001277497.1 | 3 - 4 | 431 | 93 | |
| NM_026319.3 | NP_080595.2 | 3 - 4 | 431 | 93 | ||
| GenBank mRNA | AK011690.1 | - | 3 - 4 | 402 | 93 | |
| AK014770.1 | - | 3 - 4 | 399 | 93 | ||
| AK053369.1 | - | 3 - 4 | 379 | 93 | ||
| BC005798.1 | - | 3 - 4 | 401 | 93 | ||
| BC023760.1 | - | 3 - 4 | 363 | 93 | ||
| BC049156.1 | - | 3 - 4 | 351 | 93 | ||
| BQ950148.1 | - | 3 - 4 | 402 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ift74
Genomic Map