Assay Details
| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104385.1 | 10 - 11 | 1311 | 128 | ||
| GenBank mRNA | Y08380.1 | - | 10 - 11 | 1259 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dab1
Genomic Map