Assay Details
| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177259.4 | NP_796233.2 | 10 - 11 | 1108 | 67 | |
| NR_104385.1 | 11 - 12 | 1378 | 67 | |||
| GenBank mRNA | BC138538.1 | - | 9 - 10 | 829 | 8861558 | 67 |
| BC138539.1 | - | 9 - 10 | 829 | 8861559 | 67 | |
| Y08379.1 | - | 10 - 11 | 1056 | 67 | ||
| Y08380.1 | - | 11 - 12 | 1326 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dab1
Genomic Map