Assay Details
| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177259.4 | NP_796233.2 | 7 - 8 | 907 | 66 | |
| NR_104385.1 | 7 - 8 | 907 | 66 | |||
| GenBank mRNA | BC138538.1 | - | 6 - 7 | 628 | 8861558 | 66 |
| BC138539.1 | - | 6 - 7 | 628 | 8861559 | 66 | |
| Y08379.1 | - | 7 - 8 | 855 | 66 | ||
| Y08380.1 | - | 7 - 8 | 855 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dab1
Genomic Map