Assay Details
| Gene Symbol: Mysm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177239.2 | NP_796213.2 | 15 - 16 | 1854 | 103 | |
| GenBank mRNA | AK173300.1 | - | 12 - 13 | 1596 | 103 | |
| BC150946.1 | - | 15 - 16 | 1850 | 9087857 | 103 | |
| BC151172.1 | - | 15 - 16 | 1850 | 9088061 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mysm1
Genomic Map