Assay Details
| Gene Symbol: Mysm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177239.2 | NP_796213.2 | 1 - 2 | 101 | 104 | |
| GenBank mRNA | AK043802.1 | - | 1 - 2 | 101 | 104 | |
| AK081684.1 | - | 1 - 2 | 76 | 104 | ||
| AK145221.1 | - | 1 - 2 | 76 | 104 | ||
| AK164103.1 | - | 1 - 2 | 47 | 104 | ||
| AK166402.1 | - | 1 - 2 | 76 | 104 | ||
| BC150946.1 | - | 1 - 2 | 97 | 9087857 | 104 | |
| BC151172.1 | - | 1 - 2 | 97 | 9088061 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mysm1
Genomic Map