Assay Details
| Gene Symbol: Bard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007525.3 | NP_031551.1 | 8 - 9 | 1936 | 108 | |
| GenBank mRNA | AF057157.1 | - | 8 - 9 | 1882 | 108 | |
| AK145528.1 | - | 8 - 9 | 1922 | 108 | ||
| BC055362.1 | - | 8 - 9 | 1868 | 108 | ||
| BC128370.1 | - | 8 - 9 | 1800 | 40111665 | 108 | |
| BC128371.1 | - | 8 - 9 | 1800 | 40111666 | 108 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bard1
Genomic Map