Assay Details
| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008001.4 | NP_032027.2 | 10 - 11 | 2345 | 64 | |
| GenBank mRNA | AK148151.1 | - | 10 - 11 | 2346 | 64 | |
| BC004838.1 | - | 2 - 3 | 235 | 64 | ||
| BC011462.1 | - | 10 - 11 | 2282 | 3674293 | 64 | |
| U22325.1 | - | 10 - 11 | 1944 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgd1
Genomic Map