Assay Details
| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008001.4 | NP_032027.2 | 2 - 3 | 985 | 71 | |
| GenBank mRNA | AK148151.1 | - | 2 - 3 | 986 | 71 | |
| BC011462.1 | - | 2 - 3 | 922 | 3674293 | 71 | |
| CN531569.1 | - | 1 - 2 | 98 | 71 | ||
| CN702035.1 | - | 2 - 3 | 475 | 71 | ||
| U22325.1 | - | 2 - 3 | 584 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgd1
Genomic Map