Assay Details
| Gene Symbol: Maged2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199246.1 | NP_001186175.1 | 11 - 12 | 1491 | 75 | |
| NM_030700.2 | NP_109625.1 | 11 - 12 | 1514 | 75 | ||
| GenBank mRNA | AF319976.1 | - | 10 - 11 | 1381 | 75 | |
| AJ277114.1 | - | 11 - 12 | 1457 | 75 | ||
| AK131598.1 | - | 6 - 7 | 553 | 75 | ||
| AK138441.1 | - | 11 - 12 | 1431 | 75 | ||
| AK169006.1 | - | 11 - 12 | 1463 | 75 | ||
| BC023319.1 | - | 9 - 10 | 1959 | 75 | ||
| BC058581.1 | - | 11 - 12 | 1470 | 6491028 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maged2
Genomic Map