Assay Details
| Gene Symbol: Pfkfb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008824.3 | NP_032850.1 | 10 - 11 | 1170 | 73 | |
| GenBank mRNA | AK132629.1 | - | 10 - 11 | 1171 | 73 | |
| BC132561.1 | - | 10 - 11 | 1155 | 40130838 | 73 | |
| BC132563.1 | - | 10 - 11 | 1155 | 40130840 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pfkfb1
Genomic Map