Assay Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025728.4 | NP_080004.1 | 4 - 5 | 619 | 127 | |
| NM_029160.3 | NP_083436.2 | 13 - 14 | 1460 | 127 | ||
| GenBank mRNA | AF490391.1 | - | 13 - 14 | 1442 | 127 | |
| AK014865.1 | - | 4 - 5 | 614 | 127 | ||
| AK016618.1 | - | 4 - 5 | 606 | 127 | ||
More Information
Set Membership: |
|

Spag16
Genomic Map