| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_001110202.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_001110202.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_001110203.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_001110203.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_001286386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_001286387.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_001286388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_053167.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
NM_053167.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012962.mm.1 |
TC1200001886.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_001110202.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_001110202.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_001110203.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_001110203.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_001286386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_001286387.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_001286388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_053167.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
NM_053167.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200012963.mm.1 |
TC1200001886.mm.1 |
MTA TC |