Assay Details
| Gene Symbol: Trim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286388.1 | NP_001273317.1 | 7 - 8 | 2051 | 85 | |
| NM_053167.3 | NP_444397.2 | 7 - 8 | 2051 | 85 | ||
| GenBank mRNA | AK049907.1 | - | 7 - 8 | 1765 | 85 | |
| AK129109.1 | - | 7 - 8 | 1756 | 85 | ||
More Information
Set Membership: |
|

Trim9
Genomic Map