Assay Details
| Gene Symbol: Trim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110202.1 | NP_001103672.1 | 8 - 9 | 2093 | 132 | |
| NM_001286387.1 | NP_001273316.1 | 10 - 11 | 2312 | 132 | ||
| NM_001286388.1 | NP_001273317.1 | 10 - 11 | 2315 | 132 | ||
| NM_053167.3 | NP_444397.2 | 10 - 11 | 2315 | 132 | ||
| GenBank mRNA | AK028280.1 | - | 8 - 9 | 1812 | 132 | |
| AK049907.1 | - | 10 - 11 | 2029 | 132 | ||
| AK129109.1 | - | 10 - 11 | 2020 | 132 | ||
| AK163123.1 | - | 10 - 11 | 2049 | 132 | ||
More Information
Set Membership: |
|

Trim9
Genomic Map