Assay Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026295.2 | NP_080571.2 | 10 - 11 | 2511 | 73 | |
| GenBank mRNA | AK156617.1 | - | 10 - 11 | 2540 | 73 | |
| BC052934.1 | - | 1 - 2 | 116 | 73 | ||
| BC053435.1 | - | 10 - 11 | 2511 | 6394578 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctdp1
Genomic Map