Assay Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011729.2 | NP_035859.2 | 3 - 4 | 695 | 77 | |
| GenBank mRNA | AK038822.1 | - | 3 - 4 | 524 | 77 | |
| AK137563.1 | - | 3 - 4 | 527 | 77 | ||
| AK140825.1 | - | 3 - 4 | 524 | 77 | ||
| AK160645.1 | - | 1 - 2 | 67 | 77 | ||
| AK164205.1 | - | 3 - 4 | 513 | 77 | ||
| BC033413.1 | - | 3 - 4 | 415 | 77 | ||
| BC053034.1 | - | 3 - 4 | 504 | 77 | ||
| BC060659.1 | - | 3 - 4 | 494 | 77 | ||
| BC092235.1 | - | 3 - 4 | 486 | 77 | ||
| BC141070.1 | - | 3 - 4 | 525 | 9055682 | 77 | |
| D16306.1 | - | 3 - 4 | 390 | 77 | ||
| U40796.1 | - | 3 - 4 | 591 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ercc5
Genomic Map