Assay Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026430.3 | NP_080706.1 | 2 - 3 | 123 | 149 | |
| GenBank mRNA | AF399958.1 | - | 2 - 3 | 123 | 149 | |
| AK005536.1 | - | 2 - 3 | 157 | 149 | ||
| AK075806.1 | - | 2 - 3 | 175 | 149 | ||
| AK152376.1 | - | 2 - 3 | 178 | 149 | ||
| BC037049.1 | - | 2 - 3 | 169 | 4935237 | 149 | |
More Information
Set Membership: |
|

Uxs1
Genomic Map