Assay Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190984.1 | NP_001177913.1 | 5 - 6 | 622 | 110 | |
| NM_001190985.1 | NP_001177914.1 | 4 - 5 | 840 | 110 | ||
| NM_021295.3 | NP_067270.1 | 5 - 6 | 656 | 110 | ||
| GenBank mRNA | AJ294535.1 | - | 5 - 6 | 649 | 110 | |
| AK146120.1 | - | 4 - 5 | 831 | 110 | ||
| BC038649.1 | - | 2 - 3 | 161 | 110 | ||
| BC058560.1 | - | 5 - 6 | 622 | 6315377 | 110 | |
| Y11550.1 | - | 5 - 6 | 621 | 110 | ||
| Y16518.1 | - | 4 - 5 | 800 | 110 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lancl1
Genomic Map