Assay Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286560.1 | NP_001273489.1 | 9 - 10 | 1061 | 128 | |
| NM_026168.4 | NP_080444.1 | 10 - 11 | 1170 | 128 | ||
| NM_026355.3 | NP_080631.1 | 10 - 11 | 1170 | 128 | ||
| GenBank mRNA | AK004661.1 | - | 10 - 11 | 834 | 128 | |
| AK007498.1 | - | 10 - 11 | 819 | 128 | ||
| AK008971.1 | - | 10 - 11 | 826 | 128 | ||
| AK009483.1 | - | 10 - 11 | 822 | 128 | ||
| AK010529.1 | - | 10 - 11 | 819 | 128 | ||
| AK016275.1 | - | 10 - 11 | 1171 | 128 | ||
| AK017876.1 | - | 10 - 11 | 854 | 128 | ||
| AK030045.1 | - | 10 - 11 | 820 | 128 | ||
| AK086727.1 | - | 10 - 11 | 822 | 129 | ||
| AK135968.1 | - | 7 - 8 | 507 | 128 | ||
| AK162267.1 | - | 9 - 10 | 794 | 128 | ||
| AK167391.1 | - | 10 - 11 | 823 | 128 | ||
| BC018188.1 | - | 10 - 11 | 822 | 3711066 | 128 | |
| BC026558.1 | - | 10 - 11 | 782 | 5030661 | 128 | |
| BC064749.1 | - | 10 - 11 | 883 | 30046220 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ergic2
Genomic Map