Assay Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286560.1 | NP_001273489.1 | 1 - 2 | 395 | 99 | |
| NM_026168.4 | NP_080444.1 | 1 - 2 | 395 | 99 | ||
| NM_026355.3 | NP_080631.1 | 1 - 2 | 395 | 99 | ||
| GenBank mRNA | AK004661.1 | - | 1 - 2 | 59 | 99 | |
| AK008971.1 | - | 1 - 2 | 51 | 99 | ||
| AK009483.1 | - | 1 - 2 | 47 | 99 | ||
| AK010529.1 | - | 1 - 2 | 44 | 99 | ||
| AK016275.1 | - | 1 - 2 | 396 | 99 | ||
| AK017876.1 | - | 1 - 2 | 79 | 99 | ||
| AK030045.1 | - | 1 - 2 | 45 | 99 | ||
| AK086727.1 | - | 1 - 2 | 47 | 99 | ||
| AK162267.1 | - | 1 - 2 | 128 | 99 | ||
| AK167391.1 | - | 1 - 2 | 48 | 99 | ||
| BC018188.1 | - | 1 - 2 | 47 | 3711066 | 99 | |
| BC064749.1 | - | 1 - 2 | 108 | 30046220 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004661.1, AK008971.1, AK009483.1, AK010529.1, AK016275.1, AK017876.1, AK030045.1, AK167391.1, BC018188.1, BC064749.1, NM_001286560.1, NM_026168.4, NM_026355.3 transcripts.
Set Membership: |
|

Ergic2
Genomic Map