Assay Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286560.1 | NP_001273489.1 | 8 - 9 | 953 | 92 | |
| NM_026168.4 | NP_080444.1 | 9 - 10 | 1062 | 92 | ||
| NM_026355.3 | NP_080631.1 | 9 - 10 | 1062 | 92 | ||
| GenBank mRNA | AK004661.1 | - | 9 - 10 | 726 | 92 | |
| AK007498.1 | - | 9 - 10 | 711 | 92 | ||
| AK008971.1 | - | 9 - 10 | 718 | 92 | ||
| AK009483.1 | - | 9 - 10 | 714 | 92 | ||
| AK010529.1 | - | 9 - 10 | 711 | 92 | ||
| AK016275.1 | - | 9 - 10 | 1063 | 92 | ||
| AK030045.1 | - | 9 - 10 | 712 | 92 | ||
| AK086727.1 | - | 9 - 10 | 714 | 92 | ||
| AK135968.1 | - | 6 - 7 | 399 | 92 | ||
| AK162267.1 | - | 8 - 9 | 686 | 92 | ||
| AK167391.1 | - | 9 - 10 | 715 | 92 | ||
| BC006895.1 | - | 9 - 10 | 680 | 2651322 | 92 | |
| BC018188.1 | - | 9 - 10 | 714 | 3711066 | 92 | |
| BC026558.1 | - | 9 - 10 | 674 | 5030661 | 92 | |
| BC064749.1 | - | 9 - 10 | 775 | 30046220 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ergic2
Genomic Map