Assay Details
| Gene Symbol: Anxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085383.1 | NP_001078852.1 | 10 - 11 | 931 | 105 | |
| NM_023628.2 | NP_076117.2 | 10 - 11 | 910 | 105 | ||
| GenBank mRNA | AJ401160.1 | - | 10 - 11 | 864 | 105 | |
| AK003395.1 | - | 10 - 11 | 931 | 105 | ||
| AK010128.1 | - | 10 - 11 | 911 | 105 | ||
| BC062140.1 | - | 10 - 11 | 921 | 30290956 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anxa9
Genomic Map