Assay Details
| Gene Symbol: Anxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085383.1 | NP_001078852.1 | 12 - 13 | 1107 | 77 | |
| NM_023628.2 | NP_076117.2 | 12 - 13 | 1086 | 77 | ||
| GenBank mRNA | AJ401160.1 | - | 12 - 13 | 1040 | 77 | |
| AK003395.1 | - | 12 - 13 | 1107 | 77 | ||
| AK010128.1 | - | 12 - 13 | 1087 | 77 | ||
| BC062140.1 | - | 12 - 13 | 1097 | 30290956 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343441 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092869 | BC093521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092869 | AK010128.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092869 | BC062140.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092869 | NM_023628.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092869 | AJ401160.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134559 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180100 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171761 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092869 | BC093521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092869 | AK010128.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092869 | BC062140.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092869 | NM_023628.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092869 | AJ401160.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P451482 | AK010128.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P451482 | BC062140.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P451482 | NM_023628.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P451482 | AJ401160.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P451482 | AK003395.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343404 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00702395 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04994765 | - | - |
| Whole Mouse Genome G4122A | A_51_P451482 | AK010128.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451482 | BC062140.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451482 | NM_023628.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451482 | AJ401160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451482 | AK003395.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117923 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171761 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02981055 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03050131 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02376218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03050134 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02981067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02981059 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03083525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03083526 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02981066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03083260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03083255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02933353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02563484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02895286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03083526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03024256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02604023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02981069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02457202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02839434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03083248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02903486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03024254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03083229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03024252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02600507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03024255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343350 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500142 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500149 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431554_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500140 | - | - |
| Mouse Genome 430 2.0 Array | 1431554_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1431554_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71790_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71790_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019145.mm.1 | NM_001085383.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019145.mm.1 | NM_001085383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019145.mm.1 | NM_023628.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019145.mm.1 | NM_023628.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019145.mm.1 | TC0300002473.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019147.mm.1 | NM_001085383.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019147.mm.1 | NM_001085383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019147.mm.1 | NM_023628.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019147.mm.1 | NM_023628.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019147.mm.1 | TC0300002473.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019148.mm.1 | NM_001085383.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019148.mm.1 | NM_001085383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019148.mm.1 | NM_023628.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019148.mm.1 | NM_023628.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019148.mm.1 | TC0300002473.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019146.mm.1 | NM_001085383.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019146.mm.1 | NM_001085383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019146.mm.1 | NM_023628.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019146.mm.1 | NM_023628.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019146.mm.1 | TC0300002473.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431554_PM_a_at | - | - |
| Mouse Expression Set 430 | 1431554_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431554_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431554_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431554_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431554_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 550340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1190195 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1190195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001190195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2715226 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2715226 | - | - |
| mouse-6 v1.1 (Illumina) | scl071790.1_55-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1190195 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl071790.1_55-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2715226 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2715226 | - | - |

Anxa9
Genomic Map