Assay Details
| Gene Symbol: Anxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023628.2 | NP_076117.2 | 1 - 2 | 104 | 72 | |
| GenBank mRNA | AJ401160.1 | - | 1 - 2 | 59 | 72 | |
| AK010128.1 | - | 1 - 2 | 105 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ401160.1, AK010128.1, NM_023628.2 transcripts.
Set Membership: |
|

Anxa9
Genomic Map